missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Claudin 11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10097702
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10097702 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10097702 Supplier Invitrogen™ Supplier No. 364500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

Claudin 11 Polyclonal Antibody for ELISA, IHC (P), IP, Western Blot

The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands. Claudin-11 is an oligodendrocyte-specific protein that is expressed in the tight junctions of Sertoli cells and myelin sheaths in mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 11
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Cldn11
Gene Accession No. O75508, Q60771
Gene Alias claudin 11; claudin 11 (oligodendrocyte transmembrane protein); Claudin11; claudin-11; Cldn11; hypothetical protein LOC508268; oligodendrocyte transmembrane protein; Oligodendrocyte-specific protein; OSP; OTM
Gene Symbols Cldn11
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of human OSP/Claudin-11, which differs by only one amino acid from mouse and rat sequences.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18417, 5010
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.