Learn More
Heidolph™ Clamp for Erlenmeyer Flasks
Product Code. 10242501 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
100 mL Erlenmeyer flasks
1000 mL Erlenmeyer flasks
2000 mL Erlenmeyer flasks
25 mL Erlenmeyer flasks
250 mL Erlenmeyer flasks
50 mL Erlenmeyer flasks
500 mL Erlenmeyer flasks
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
10242501 25 mL Erlenmeyer flasks Each
10739171 50 mL Erlenmeyer flasks Each
10749171 100 mL Erlenmeyer flasks Each
10151361 250 mL Erlenmeyer flasks Each
10789551 500 mL Erlenmeyer flasks Each
10099340 1000 mL Erlenmeyer flasks Each
11711659 2000 mL Erlenmeyer flasks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10242501 Supplier Heidolph™ Supplier No. 5495100000
Only null left
Add to basket
Add to basket
Clamps for use with various perforated platforms
  • Maximum number of clamps per platform type:
    • 549-51000-00
      • For platform 100 - max. 16
      • For platform 1000 - max. 20
      • For platform 2000 - max. 36
    • 549-52000-00
      • For platform 100 - max. 16
      • For platform 1000 - max. 20
      • For platform 2000 - max. 36
    • 549-55000-00
      • For platform 100 - max. 3
      • For platform 1000 - max. 4
      • For platform 2000 - max. 9
    • 549-56000-00
      • For platform 100 - max. 2
      • For platform 1000 - max. 4
      • For platform 2000 - max. 5
    • 549-63000-00
      • For platform 1000 - max. 2
      • For platform 2000 - max. 3
TRUSTED_SUSTAINABILITY
Specifications
Holds 25 mL Erlenmeyer flasks
Product Type Clamp
For Use With (Equipment) Perforated platforms 100, 1000 and 2000
Size 1