missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CKS1 Monoclonal Antibody (4G12G7)
Description
CKS1 Monoclonal Antibody for Western Blot, IHC, ELISA
CKS1B protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS1B mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects a specialized role for the encoded protein. At least two transcript variants have been identified for this gene, and it appears that only one of them encodes a protein.
Specifications
Specifications
| Antigen | CKS1 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 4G12G7 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | CKS1B |
| Gene Accession No. | P61024 |
| Gene Alias | 2410005G18Rik; 2610005D03Rik; AA407784; CDC28 protein kinase 1; CDC28 protein kinase 1B; CDC28 protein kinase regulatory subunit 1; CDC28 protein kinase regulatory subunit 1B; CDC2-associated protein CKS1; cell division control protein CKS1; CKS1; CKS-1; Cks1b; ckshs1; cyclin-dependent kinase regulatory subunit 1; cyclin-dependent kinases regulatory subunit 1; NB4 apoptosis/differentiation related protein; PNAS-143; PNAS-16; PNAS-18; RGD1561797; Sid 1334; sid1334; Unknown (protein for MGC:142665); zgc:101026 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?