missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trisodium salt, 98%, pure, anhydrous

Product Code. 10045303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045303 2.5 kg Plastic bottle 2.5kg
10362234 100 g Plastic bottle 100g
10360334 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10045303 Supplier Thermo Scientific Acros Supplier No. 391970025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68-04-2 | C6H5Na3O7 | 258.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-04-2
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00012462
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Synonym sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous
PubChem CID 6224
ChEBI CHEBI:53258
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Melting Point 300.0°C
Color White
Flash Point 290°C
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Plastic bottle
Quantity 2.5 kg
Merck Index 15, 8737
Solubility Information Solubility in water: 500g/L (20°C).
Formula Weight 258.07
Percent Purity 0.98
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Citric acid, trisodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.