missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl bromide, 97%, predominantly trans

Product Code. 10041781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041781 100 g Glass bottle 100g
10787461 5 g Glass bottle 5g
10122261 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10041781 Supplier Thermo Scientific Acros Supplier No. 110401000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4392-24-9 | C9H9Br | 197.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4392-24-9
Molecular Formula C9H9Br
Molecular Weight (g/mol) 197.07
MDL Number MFCD00000245
InChI Key RUROFEVDCUGKHD-QPJJXVBHSA-N
Synonym cinnamyl bromide, 3-bromoprop-1-en-1-yl benzene, e-3-bromoprop-1-en-1-yl benzene, 3-bromo-1-phenyl-1-propene, 3-bromo-1-phenyl propene-1, e-3-bromoprop-1-enyl benzene, 3-bromo-propenyl-benzene, e-cinnamyl bromide, trans-cinnamyl bromide
PubChem CID 5357478
IUPAC Name [(E)-3-bromoprop-1-enyl]benzene
SMILES C1=CC=C(C=C1)C=CCBr

Specifications

Melting Point 26°C to 32°C
Density 1.3320g/mL
Boiling Point 103°C (22.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCH2Br
Refractive Index 1.621
Quantity 100 g
Beilstein 05, II, 372
Specific Gravity 1.332
Formula Weight 197.07
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Cinnamyl bromide, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.