missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CHUK (IKK Alpha), His Tag Recombinant Protein

Product Code. 10056782
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10056782 1 mg 1mg
15468613 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10056782 Supplier Invitrogen™ Supplier No. PV4327

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length

CHUK (IKK alpha) is a serine/threonine kinase involved in signal transduction and cellular communication. It belongs to the cytokine activated protein complex that is an inhibitor of the NF-kappa B complex. IKK alpha along with IKK beta and a regulatory protein NEMO (IKK gamma) phosphorylates IkB proteins which targets the later for ubiquitin mediated degradation. It is involved with several cellular processes with extensive studies on its role in inflammation and cell survival.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15111
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1147
Molecular Weight (g/mol) 88.7 kDa
Name Human CHUK (IKK Alpha), His Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Gene Alias AI256658; CHUK; Chuk1; component of inhibitor of nuclear factor kappa B kinase complex; conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; Fbx24; Fbxo24; FLJ40509; I kappa B kinase 1; i kappa-B kinase alpha; ikappaB kinase; I-kappa-B kinase 1; I-kappa-B kinase 2; IkappaB kinase alpha; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; IkB kinase alpha subunit; IkB kinase-alpha; ikBKA; IKBKB; IKK alpha; IKK1; IKK2; Ikka; IKK-A; IKK-a kinase; IKK-alpha; IKKB; IKK-B; IKK-beta; Inhibitor of nuclear factor kappa-B kinase subunit alpha; kinase beta; MGC131801; NFKBIKA; NFKBIKB; Nuclear factor NFkappaB inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase alpha; TCF16; TCF-16; Transcription factor 16
Common Name IKK alpha
KinaseFamily IKK Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.