missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chromoxane Cyanine R, pure

Product Code. 10012670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10012670 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10012670 Supplier Thermo Scientific Chemicals Supplier No. 215150050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3564-18-9 | C23H15Na3O9S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

CAS 3564-18-9
Molecular Formula C23H15Na3O9S
MDL Number MFCD00012411
Beilstein 11, II, 240
Synonym C.I. 43820, Mordant Blue 3
Infrared Spectrum Authentic
Formula Weight 536.39
Grade Extra Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Brown to Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Chromoxane Cyanine R
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.