missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromate Standard: CrO4(-2) @ 10000 μg/mL in H2O, VHG™
Description
Chromate Standard: CrO4(-2) @ 10000 μg/mL in H2O
Specifications
Specifications
| Chemical Name or Material | Chromate |
| CAS | 11104-59-9 |
| Methods | Ion Chromatography/Wet Chemistry |
| Analyte or Component Names | Chromate |
| Shipping Condition | Room Temperature |
| Molecular Formula | (Cr O4)2- |
| Synonym | CrO42− |
| SMILES | [O-][Cr](=O)(=O)[O-] |
| IUPAC Name | Chromate |
| Solvent or Matrix | Water |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?