missing translation for 'onlineSavingsMsg'
Learn More

Cholic acid, MedChemExpress

Product Code. p-200097296
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452519 1 g 1g
16419056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452519 Supplier Medchem Express Supplier No. HYN0324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cholic acid is a major primary bile acid produced in the liver and usually conjugated with glycine or taurine. It facilitates fat absorption and cholesterol excretion. Cholic acid is orally active.

Cholic acid is a major primary bile acid produced in the liver and usually conjugated with glycine or taurine. It facilitates fat absorption and cholesterol excretion. Cholic acid is orally active.

Specifications

Chemical Name or Material Cholic acid
CAS 81-25-4
Color White
Purity Grade Notes Research
Molecular Formula C24H40O5
Quantity 1 g
Solubility Information DMSO : ≥ 50 mg/mL (122.38 mM) ∣H2O : 0.67 mg/mL (1.64 mM; Need ultrasonic)
SMILES C[C@H](CCC(O)=O)[C@@]1([H])CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@@]21C
Molecular Weight (g/mol) 408.57
Formula Weight 408.57
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.