missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl benzoate, 98%

Product Code. 10045923 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045923 100 g Glass bottle 100g
10132280 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10045923 Supplier Thermo Scientific Acros Supplier No. 110201000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 604-32-0 | C34H50O2 | 490.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-32-0
Molecular Formula C34H50O2
Molecular Weight (g/mol) 490.77
MDL Number MFCD00003635
InChI Key UVZUFUGNHDDLRQ-UHFFFAOYNA-N
Synonym cholesteryl benzoate, cholesterol benzoate, 3b-benzoyloxy cholest-5-ene, unii-n09h13shlb, n09h13shlb, cholest-5-ene-3-beta-yl benzoate, cholest-5-en-3-ol 3.beta.-, benzoate, 5-cholesten-3.beta.-ol benzoate, cholest-5-en-3-ol 3beta-, benzoate
PubChem CID 2723613
SMILES CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(=O)C1=CC=CC=C1

Specifications

Melting Point 149°C to 150°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 10, 2181
Specific Rotation -15
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,methylene chloride,,benzene,toluene,diethyl ether
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] benzoate
Formula Weight 490.76
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Cholesteryl benzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.