missing translation for 'onlineSavingsMsg'
Learn More

Chloroprene 5000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30311304
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30311304 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30311304

Brand: Dr. Ehrenstorfer DREGA09010381ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloroprene
CAS 126-99-8
Analyte or Component Names Chloroprene
Shipping Condition Ice pack (-20°C)
Molecular Formula C4 H5 Cl
Concentration 5000 μg/mL
Certifications/Compliance ISO 17034
Synonym 1,3-Butadiene, 2-chloro-, 2-Chloro-1,3-butadiene, 2-Chloro-1,3-buatdiene, 2-Chlorobutadiene, Chloroprene, NSC 18589, β-Chloroprene
InChI Formula InChI=1S/C4H5Cl/c1-3-4(2)5/h3H,1-2H2
SMILES ClC(=C)C=C
IUPAC Name 2-chlorobuta-1,3-diene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 88.54
Quantity 1 mL
Formula Weight 88.008 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.