missing translation for 'onlineSavingsMsg'
Learn More

Chloroform-13C, TRC

Product Code. 30354497
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354497 Supplier Toronto Research Chemicals Supplier No. TRCC3665031G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloroform 13C
CAS 31717-44-9
Molecular Formula 13C H Cl3
Quantity 1 g
Synonym Trichloromethane 13C
InChI Formula InChI=1S/CHCl3/c2-1(3)4/h1H/i1+1
SMILES Cl[13CH](Cl)Cl
IUPAC Name trichloro(1^{13}C)methane
Molecular Weight (g/mol) 120.37
Formula Weight 118.918
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.