missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(cyclooctene)rhodium(I) dimer, 98%

Product Code. 10008881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10008881 100 mg Glass bottle 100mg
10602233 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10008881 Supplier Thermo Scientific Acros Supplier No. 369371000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12279-09-3 | C32H56Cl2Rh2 | 717.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12279-09-3
Molecular Formula C32H56Cl2Rh2
Molecular Weight (g/mol) 717.51
MDL Number MFCD00013287
InChI Key GQPAPAIPOLEZHT-XFCUKONHSA-L
Synonym chlorobis cyclooctene rhodium i dimer, chlorobis cyclooctene rhodium dimer
PubChem CID 53384308
IUPAC Name cyclooctene;rhodium;dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1

Specifications

Melting Point 187.0°C to 191.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 717.52
Percent Purity 98%
Chemical Name or Material Chlorobis(cyclooctene)rhodium(I) dimer
compliance-icons
Product Identifier
  • Chlorobis(cyclooctene)rhodium(I) dimer
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.