missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chiller Plumbing Kits and Accessories

Product Code. 15176445 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Fittings
Plumbing Package
Tubing
Tubing Kit
For Use With (Equipment):
Merlin Chillers with PD1/PD2 pump
ThermoFlex 3500 to 5000
ThermoFlex 900 to 10000
ThermoTemp, Polar
ThermoTemp, Polar, VersaCool
Size:
1/2 in. NPT, 25 ft.
1/2 in. NPT, 50 ft.
1/2 to 3/8 in. NPT
3/4 in. NPT, 32.8 ft.
6 mm ID per meter, 0.5 meter long
6 mm ID per meter, 1.0 m
6 mm ID per meter, 1.0 meter long
6 mm ID per meter, 1.5 m
6 mm ID per meter, 1.5 meter long
8 mm ID per meter
Material:
Brass
Flexible Plastic
Perbunan
Polybraid
Stainless-steel
Unit Size:
1m
Each
Pack of 2
Product Code. Type For Use With (Equipment) Size Material unitSize
15176445 Plumbing Package Merlin Chillers with PD1/PD2 pump 1/2 in. NPT, 25 ft. Flexible Plastic Each
17108165 Plumbing Package ThermoFlex 3500 to 5000 3/4 in. NPT, 32.8 ft. Polybraid Each
11957514 Tubing Kit ThermoFlex 900 to 10000 1/2 in. NPT, 50 ft. Polybraid Each
10430463 Tubing ThermoTemp, Polar, VersaCool 6 mm ID per meter, 1.0 meter long Stainless-steel Each
10505872 Tubing ThermoTemp, Polar 6 mm ID per meter, 1.0 m Stainless-steel Each
10771993 Tubing ThermoTemp, Polar 6 mm ID per meter, 1.5 m Stainless-steel Each
10719181 Tubing ThermoTemp, Polar, VersaCool 6 mm ID per meter, 0.5 meter long Stainless-steel Each
10111031 Tubing ThermoTemp, Polar, VersaCool 6 mm ID per meter, 1.5 meter long Stainless-steel Each
10258621 Tubing ThermoTemp, Polar 8 mm ID per meter Perbunan 1m
15865038 Fittings ThermoFlex 900 to 10000 1/2 to 3/8 in. NPT Brass Pack of 2
10 options
This item is not returnable. View return policy

Product Code. 15176445

Brand: Thermo Scientific™ 611000000012

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.

Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.

Features:

  • Include flexible tubing, tubing insulation, and hose clamps
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Merlin Chillers with PD1/PD2 pump
Type Plumbing Package
Includes 25 ft. of 1/2 in. ID flexible tubing, 25 ft. of tubing insulation, and hose clamp
Material Flexible Plastic
Size 1/2 in. NPT, 25 ft.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.