Learn More
Aqualytic™ Checkit Ozone DPD Test Kit
Product Code. 10691433
Change view
Click to view available options
Range:
0 to 1.0mg/L
Unit Size:
Each
Product Code. Range unitSize
10691433 0 to 1.0mg/L Each
1 options
Product Code. 10691433 Supplier Aqualytic™ Supplier No. 147270
Aqualytic Checkit Ozone DPD Test Kit, Range: 0 to 1.0mg/L, Accuracy: +/-5%, Quantity: 1Pack
Specifications
Accuracy ±5%
Quantity 1 Pack
Range 0 to 1.0mg/L
Type Checkit Ozone DPD Test Kit
compliance-icons
Product Identifier
  • COMPARATOR LOVIBOND 147270 OZONE 0 TO 0.7MG/L .
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide