Learn More
Aqualytic™ Checkit™ 5 in 1 Test Kit
Product Code. 10208204
Change view
Click to view available options
Quantity:
1 Pack
Unit Size:
Each
Product Code. Quantity unitSize
10208204 1 Pack Each
1 options
Product Code. 10208204 Supplier Aqualytic™ Supplier No. 147028
Aqualytic™ Checkit™ 5 in 1 Test Kit includes all materials and operating instructions needed for mobile and static water analysis of 5 different components and comes in a sturdy and handy plastic case.
  • Tests for chlorine, ph, cyanuric acid (turbidity method), calcium hardness (speed test), and total alkalinity (speed test)
  • CHECKIT™ Comparator
    • Compact, handy colorimetric unit suitable for mobile and stationary analyses
    • Supplied with a generous number of different color scales
  • CHECKIT™ Discs
    • Continuous color scale makes it possible to achieve an exact color match between the standard and the sample.
    • Specially manufactured in selected materials to remain color stable over a long period of time for reliable, reproducible measurement results
  • AQUALYTIC™ tablet reagents (for 30 tests)
  • Stirring Rod
  • Cells
  • Operating instructions
    • Step-by-step explanation of how to conduct the test, ensuring reliable and fast and accurate measurements
Specifications
Includes Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions
No. of Tests 30
Quantity 1 Pack
Range 6.5 to 8.4pH, 0 to 4.0mg/L Cl2, 20 to 200mg/L Cys, 20 to 800mg/L CaCO3, 20 to 800mg/L CaCO3
Sufficient For 30 Tests
Test Method Turbidity, speed test
Type Checkit™ 5 in 1 Test Kit