missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CGP 20712 dihydrochloride
GREENER_CHOICE

Product Code. 30001162
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001162 10 mg 10mg
18025152 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001162 Supplier Tocris Bioscience™ Supplier No. 1024/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent and selective β1 antagonist

CGP 20712 dihydrochloride is a highly selective and potent β1-adrenoceptor antagonist (IC50 = 0.7 nM). Displays 10,000-fold selectivity over β2-adrenoceptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216905-73-5
Molecular Formula C23H27Cl2F3N4O5

Specifications

Quantity 10 mg
Formula Weight 567.39
Purity >96%
Molecular Weight (g/mol) 567.387
Chemical Name or Material CGP 20712 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.