missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetyl Trimethylammonium Bromide, Certified AR for Analysis, Fisher Chemical™
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Melting Point | 230°C |
| Color | White |
| pH | 5 to 7 |
| Physical Form | Powder/Solid |
| Quantity | 500 g |
| Formula Weight | 364.45g/mol |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Hexadecyltrimethylammonium bromide
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?