missing translation for 'onlineSavingsMsg'
Learn More

Cesium Hydroxide Monohydrate, Honeywell Fluka™

Product Code. p-7071865 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15611910 10 g 10g
15631910 50 g 50g
15621910 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15611910 Supplier Honeywell-Fluka Supplier No. 2100010G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥95.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35103-79-8
Molecular Formula CsH3O2
Molecular Weight (g/mol) 167.93
MDL Number MFCD00010964
InChI Key ABSOMGPQFXJESQ-UHFFFAOYSA-M
Synonym cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
PubChem CID 23679066
SMILES O.[OH-].[Cs+]

Specifications

Quantity 10 g
UN Number 2682
IUPAC Name cesium;hydroxide;hydrate
Formula Weight 167.93g/mol
Percent Purity ≥95% (T)
Density 3.68g/mL at 25°C
Chemical Name or Material Cesium Hydroxide Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.