missing translation for 'onlineSavingsMsg'
Learn More

Cesium acetate, 99%, for analysis

Product Code. 10186863 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10186863 25 g Glass bottle 25g
10433195 100 g Glass bottle 100g
10196863 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10186863 Supplier Thermo Scientific Acros Supplier No. 390830250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3396-11-0 | C2H3CsO2 | 191.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.96
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Melting Point 194.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 25 g
Assay Percent Range 99%
Linear Formula CH3CO2Cs
Solubility Information Solubility in water: 91.5% (21°C).
Formula Weight 191.96
Percent Purity 99%
Grade Analytical
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Cesium acetate
Show More Show Less
compliance-icons
Product Identifier
  • Cesium acetate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid, cesiumsalt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.