missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Ceramus™ Fix Volume Bottle Top Dispensers
Description
- For fixed and variable volumes, from 0.2 mL to 60 mL
- Complete unit can be steam sterilized at 121°C without dismantling
- Free rotation
- Media recirculating system for simple and rapid venting without reagent loss
- Digital volume configuration engagement, calibrated for a lifetime
- Use within a temperature range of 4°C to 50°C
- Individual quality certificate with conformity certificate
Specifications
Specifications
| Accuracy | 0.6% |
| Product Type | Bottletop Dispenser |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO 8655 |
| Description | ceramus™ FIX, fixed volume bottle top dispenser with reagent recirculation system |
| Material | Aluminum Oxide Ceramic Piston, Duran Glass Cylinder |
| Volume (Metric) | 2 fix |
| For Use With (Equipment) | Bottle |
| Includes | Thread adapter, dispensing nozzle, suction tube, valve wrench, calibration report |
| Increments | 0.05 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?