missing translation for 'onlineSavingsMsg'
Learn More

Cellulose microcrystalline, average particle size 50 μm

Product Code. 10166663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10166663 1 kg Plastic Bottle 1kg
10065563 250 g Plastic Bottle 250g
11926991 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10166663 Supplier Thermo Scientific Acros Supplier No. 387240010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9004-34-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6
MDL Number MFCD00081512

Specifications

Color White
pH 5 to 7
Physical Form Micro Fine Crystalline Powder
Linear Formula (C12H20O10)n
Solubility Information Solubility in water: insoluble.
Quantity 1 kg
Infrared Spectrum Authentic
Loss on Drying 1.5% max.
Packaging Plastic Bottle
Particle Size 250 Micron <1% min., 75 Micron <30% min., 32 Micron >50% min.
Residue after Ignition 0.05% max.
Chemical Name or Material Cellulose microcrystalline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.