missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDKN2C Monoclonal Antibody (ZP001)
GREENER_CHOICE

Product Code. 10139352
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10139352 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10139352 Supplier Invitrogen™ Supplier No. 393400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

CDKN2C Monoclonal Antibody for Western Blot, ICC/IF, ELISA

The protein encoded by this gene is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to interact with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. Ectopic expression of this gene was shown to suppress the growth of human cells in a manner that appears to correlate with the presence of a wild-type RB1 function. Studies in the knockout mice suggested the roles of this gene in regulating spermatogenesis, as well as in suppressing tumorigenesis. Two alternatively spliced transcript variants of this gene, which encode an identical protein, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKN2C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ZP001
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CDKN2C
Gene Accession No. P42773, Q60772
Gene Alias C77269; CDK6 inhibitor p18; Cdkn2c; CDKN6; cyclin dependent kinase inhibitor 2C; cyclin-dependent inhibitor; cyclin-dependent kinase 4 inhibitor C; Cyclin-dependent kinase 6 inhibitor; cyclin-dependent kinase 6 inhibitor p18; cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4); INK4c; p18; p18INK4c; p18-INK4c; p18-INK6
Gene Symbols CDKN2C
Host Species Mouse
Immunogen Recombinant, full-length mouse p18INK4c.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1031, 12580
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.