missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD79b Monoclonal Antibody (SN8), FITC

Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
Specifications
Specifications
| Antigen | CD79b |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SN8 |
| Concentration | 0.1mg/mL |
| Conjugate | FITC |
| Description | CD79b Monoclonal Antibody, 0.5mL, 0.1mg/mL Concentration, SN8 Clone, CD79b Antigen, Human CD79b Immunogen, Mouse Host, Human Reactivity, FITC Conjugate, IgG1, kappa Isotype, Used For Flow Cytometry |
| Dilution | Assay-dependent |
| Formulation | PBS with BSA and 0.1% sodium azide |
| Gene | Cd79b |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?