missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD18 Monoclonal Antibody (MEM-48)
GREENER_CHOICE

Product Code. 10027282
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10027282 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10027282 Supplier Invitrogen™ Supplier No. 371300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

CD18 Monoclonal Antibody for Western Blot, ICC/IF

CD18, also known as the integrin beta 2 subunit, is a 90-95 kDa type I transmembrane protein expressed on all leukocytes. It heterodimerizes with the alpha chains of CD11a, CD11b, CD11c, and CD11d to form various leukocyte (beta2) integrins, which are crucial for cellular adhesion and signaling. The CD18/CD11a complex, known as lymphocyte function-associated antigen-1 (LFA-1), plays a significant role in cellular adhesion and inflammatory reactions. CD18 also forms heterodimers with CD11b (Mac-1, CR3), CD11c, and CD11d, contributing to the processes of cell adhesion and cell-surface mediated signaling. These integrins are essential for proper leukocyte migration and mediating intercellular contacts. The absence of CD18 leads to leukocyte adhesion deficiency type I (LAD1), a condition characterized by impaired leukocyte migration. Severe reduction in CD18 expression can result in the development of a psoriasiform skin disease. CD18 is also a target of Mannheimia (Pasteurella) haemolytica leukotoxin, which can mediate leukotoxin-induced cytolysis. Defects in the CD18 gene are the cause of LAD1, and two transcript variants encoding the CD18 protein have been identified. The integrin-mediated responses are often a composite of the functions of its individual subunits, highlighting the importance of CD18 in immune function and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MEM-48
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene ITGB2
Gene Accession No. P05107
Gene Alias 2E6; AI528527; antigen CD18; B-2 integrin; CD18; CD18 leukocyte adhesion molecule; CD18 subunit; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); cell surface adhesion glycoproteins LFA-1/CR3/p150,95 subunit beta; complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; complement receptor C3 subunit beta; integrin beta 2; integrin beta chain, beta 2; integrin beta-2; integrin subunit beta 2; integrin, beta 2; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; macrophage antigen 1 (mac-1) beta subunit); integrin, beta 2 (antigen CD18 subunit (p95), lymphocyte function-associated antigen 1, integrin B2); integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); ITGB2; LAD; LCAMB; Leu-CAM receptor; leukocyte cell adhesion molecule CD18; leukocyte surface protein; leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C; Lfa1; LFA-1; lymphocyte function associated antigen 1; MAC-1; Mac-1 beta; macrophage antigen 1 (mac-1) beta subunit); macrophage antigen-1 beta; MF17; MFI7
Gene Symbols ITGB2
Host Species Mouse
Immunogen Human CD18.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.