missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD14 Monoclonal Antibody (TuK4)

Product Code. 10038582
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10038582 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10038582 Supplier Invitrogen™ Supplier No. MHCD1400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

CD14 Monoclonal Antibody for Flow

CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD14
Applications Flow Cytometry
Classification Monoclonal
Clone TuK4, TüK4
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Cd14
Gene Accession No. P08571
Gene Alias CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14
Gene Symbols Cd14
Host Species Mouse
Immunogen Human CD14.
Purification Method Purified
Quantity 500 μL
Regulatory Status ASR, ASR/IVD
Primary or Secondary Primary
Gene ID (Entrez) 929
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.