missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD105 Monoclonal Antibody (SN6)
GREENER_CHOICE

Product Code. 10038452
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10038452 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10038452 Supplier Invitrogen™ Supplier No. MHCD10500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

CD105 Monoclonal Antibody for Flow

CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD105
Applications Flow Cytometry
Classification Monoclonal
Clone SN6
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ENG
Gene Accession No. P17813
Gene Alias AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein
Gene Symbols ENG
Host Species Mouse
Immunogen Human CD105 (Endoglin).
Purification Method Purified
Quantity 500 μL
Regulatory Status ASR
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.