missing translation for 'onlineSavingsMsg'
Learn More

CC-115, MedChemExpress

Product Code. p-200100435
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499747 5 mg 5mg
16417228 10 mg 10mg
16413247 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499747 Supplier Medchem Express Supplier No. HY16962

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CC-115 is a potent and dual DNA-PK and mTOR kinase inhibitor with IC50s of 13 nM and 21 nM, respectively. CC-115 blocks both mTORC1 and mTORC2 signaling.

CC-115 is a potent and dual DNA-PK and mTOR kinase inhibitor with IC50s of 13 nM and 21 nM, respectively. CC-115 blocks both mTORC1 and mTORC2 signaling.

Specifications

Chemical Name or Material CC-115
CAS 1228013-15-7
Color Red
Purity Grade Notes Research
Molecular Formula C16H16N8O
Quantity 5 mg
Solubility Information DMSO : 20 mg/mL (59.46 mM; Need ultrasonic)
SMILES O=C1CN=C2C(N1CC)=NC(C3=C(C)N=C(C4=NN=CN4)C=C3)=CN2
Molecular Weight (g/mol) 336.35
Formula Weight 336.35
Percent Purity 95.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.