missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps
GREENER_CHOICE

Product Code. 17105311
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
No. per Case:
2000/Cs.
Unit Size:
Pack of 2000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color No. per Case unitSize
17105311 Red 2000/Cs. Pack of 2000
17115311 White 2000/Cs. Pack of 2000
17125311 Blue 2000/Cs. Pack of 2000
17145311 Yellow 2000/Cs. Pack of 2000
17135311 Green 2000/Cs. Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17105311 Supplier Thermo Scientific™ Supplier No. S40200
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps are made of polyethylene.

Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps are made of polyethylene.

  • For 10 and 12 mL serum transport tubes
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outer 15 mm
Product Type Screw Cap
Material Polyethylene
Color Red
Height (Metric) 96 mm
No. per Case 2000/Cs.
For Use With (Equipment) Capitol Vial 10 and 12 mL Transport Tubes (not included)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.