Learn More
Trajan Scientific and Medical™ Capillary Column Mini Unions
Product Code. 10095320
Change view
Click to view available options
Diameter (Metric) Inner:
0.3 mm
0.4 mm
0.5 mm
0.8 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10095320 0.5 mm Each
10516772 0.3 mm Each
10310471 0.4 mm Each
10686971 0.8 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10095320 Supplier Trajan Scientific and Medical™ Supplier No. 103432
Only null left
Add to basket
Add to basket
SilTite Mini Union delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Type Capillary Column Connector
Material Stainless Steel
Diameter (Metric) Inner 0.5 mm