Learn More
Invitrogen™ Human CAMKK2 (CaMKK Beta), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | Q96RR4 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 10645 |
| Molecular Weight (g/mol) | 87.1 kDa |
| Name | Human CAMKK2 (CaMKK Beta), GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | 6330570N16Rik; AW061083; Ca+/Calmodulin-dependent protein kinase kinase beta (CaM-kinase kinase beta); calcium/calmodulin dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase 2 beta; calcium/calmodulin-dependent protein kinase beta; calcium/calmodulin-dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase kinase 2, beta; calcium/calmodulin-dependent protein kinase kinase beta; caM-kinase kinase 2; CaM-kinase kinase beta; CAMKK; caMKK 2; caM-KK 2; caMKK beta; CaM-KK beta; CAMKK beta protein; CAMKK2; CAMKKB; KIAA0787; MGC15254; mKIAA0787 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.