missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CAMK1D (CaMKI Delta), His Tag Recombinant Protein

Product Code. 10086822
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10086822 1 mg 1mg
15408493 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10086822 Supplier Invitrogen™ Supplier No. PV3977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

CAMK1D (CaMK1 delta) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. CAMK1D is expressed in polymorphonuclear leukocytes and may be part of the chemokine signal transduction pathway that regulates granulocyte function. It may also be involved in modulation of neuronal apoptosis. Calcium/clamodulin-dependent protein kinase 1D-beta isoform is a novel calmodulin-kinase that exhibits Ca (2+)/CaM-dependent activity. CAM-Kkalpha upregulates CAMK1D-b activity through phosphorylation of its Thr180.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8IU85
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 57118
Molecular Weight (g/mol) 47 kDa
Name Human CAMK1D (CaMKI Delta), His Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Gene Alias A630059D12Rik; calcium/calmodulin dependent protein kinase ID; calcium/calmodulin-dependent protein kinase 1 D; calcium/calmodulin-dependent protein kinase ID; calcium/calmodulin-dependent protein kinase type 1 D; CaM kinase I delta; caM kinase ID; CaM-K1; CAMK1D; CAMKI; caMKI delta; caM-KI delta; CAMKID; CaMKIdelta; CaMKI-delta; camKI-like protein kinase; CKLiK; E030025C11Rik; mCKLiK; MGC120317; MGC120318; OTTHUMP00000019119; OTTHUMP00000019120; OTTHUMP00000045030; RGD1560691; RP11-462F15.1
Common Name CAMK1D
KinaseFamily CAMK1 Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.