missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate hemihydrate, 97+%, pure

Product Code. 10075243 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10075243 2.5 kg Plastic bottle 2.5kg
10298653 100 g Plastic bottle 100g
10367583 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10075243 Supplier Thermo Scientific Acros Supplier No. 385350025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10034-76-1 | 0·5 H2O | 145.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-76-1
Molecular Formula 0·5 H2O
Molecular Weight (g/mol) 145.15
MDL Number MFCD00149624
InChI Key ZOMBKNNSYQHRCA-UHFFFAOYSA-J
Synonym calcium sulfate hemihydrate, gypsum hemihydrate, calcium sulphate hemihydrate, unii-3rw091j48v, crystacal, densite, hemihydrate gypsum, densite gypsum, gipshalbhydrat, p 23 sulfate
PubChem CID 3033839
ChEBI CHEBI:32584
IUPAC Name dicalcium;disulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 97% min. (On dry substance) (Complexometry)
Solubility Information Solubility in water: 0.8g/L (20°C)
Formula Weight 145.15
Percent Purity ≥97%
Grade Pure
Identification Confirmed
Loss on Ignition 4.5% to 8.0% (500°C, 2 Hrs)
Packaging Plastic bottle
Chemical Name or Material Calcium sulfate hemihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.