missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, anhydrous, J.T. Baker™

Product Code. 10162400 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10162400 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10162400 Supplier JT Baker Supplier No. 0070.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laboratory Reagent

Specifications

Boiling Point 1600°C (1013 hPa)
Melting Point 772°C
Quantity 1 kg
Assay Percent Range min. 95%
Molecular Formula CaCl2
MDL Number MFCD00010903
Molecular Weight (g/mol) 110.98
Packaging Plastic bottle
Density 2.174 g/cm³ (20°C)
compliance-icons
Product Identifier
  • Calcium Chloride Anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Calcium Chloride Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.