Learn More
Uvex™ C300 Dry Cut Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
8
Unit Size:
Each
Description
- Gloves provide level 3/C blade cut resistance, and very high tear resistance
- High-performance vinyl (HPV) dots on the palm enhance grip capabilities in dry environments
- Uvex Climazone technology minimizes excessive warmth and moisture
- Highly flexible with very good tactile sensation
- Silicone-free according to the imprint test
- 3D Ergo technology ensures a secure fit
Specifications
Specifications
| Coating Material | HPV Dots |
| Color | Gray, Green |
| Material | Bamboo Viscose, HPPE, Polyamide, Glass |
| Size | 8 |
| EN388 Standard | XX4XC |
| Glove Cuff Style | Knit |
| Series | C300 Dry |