missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Jun Monoclonal Antibody (ZC008)
GREENER_CHOICE

Product Code. 10149352
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10149352 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10149352 Supplier Invitrogen™ Supplier No. 397500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

c-Jun Monoclonal Antibody for Western Blot, ICC/IF, ELISA

c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Jun
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ZC008
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene JUN
Gene Accession No. P05412, P05627
Gene Alias Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Gene Symbols JUN
Host Species Mouse
Immunogen Recombinant protein encompassing an internal region of the human c-Jun protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008856, 16476, 3725
Target Species Human, Mouse, Rabbit
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.