missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ C 176

Description
C 176 is a mSTING antagonist. Covalently binds Cys91 of mouse but not human STING. Blocks activation-induced palmitoylation of STING and formation of STING clusters. Reduces CMA (Cat. No. 6677) -induced increase in serum IFNs in vivo and suppresses systemic inflammation in a mouse inflammatory disease model.
Specifications
Specifications
| CAS | 314054-00-7 |
| Quantity | 50 mg |
| Target | STING Antagonists |
| Molecular Formula | C11H7IN2O4 |
| Formula Weight | 2929 |
| Purity | 97% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?