missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Busulfan-d8 (tetramethylene-d8), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Busulfan-d8 |
| CAS | 116653-28-2 |
| Molecular Formula | C6 2H8 H6 O6 S2 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C6H14O6S2/c1-13(7,8)11-5-3-4-6-12-14(2,9)10/h3-6H2,1-2H3/i3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])(OS(=O)(=O)C)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OS(=O)(=O)C |
| IUPAC Name | (1,1,2,2,3,3,4,4-octadeuterio-4-methylsulfonyloxybutyl) methanesulfonate |
| Molecular Weight (g/mol) | 254.35 |
| Formula Weight | 254.0734 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction