Learn More
Lenz Laborglasintrumente Burette Stopcock, Lateral, with Glass Plug
Product Code. 10090351
Change view
Click to view available options
:
NS 12.5
Product Code.
10090351 NS 12.5
1 options
Product Code. 10090351 Supplier Lenz Laborglasintrumente Supplier No. 02702001
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Facetted tip
  • Bore: 1.5 mm
  • Stopcock: NS 12.5
  • Solid NS glass plug
  • Screwthread retaining device
  • Material: borosilicate glass 3.3
Specifications
Material Borosilicate Glass (3.3)
Stopcock NS 12.5