missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 7, Phosphate buffer

Product Code. 10114680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10114680 1 L 1 L
10448273 100 mL 100mL
10317531 500 mL 500mL
11906971 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10114680 Supplier Thermo Scientific Acros Supplier No. 258590010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7558-79-4 | H2KO4P

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 7
CAS 7558-79-4, 7732-18-5
Quantity 1 L
Molecular Formula H2KO4P
MDL Number MFCD03097944
Packaging Plastic bottle
Merck Index 14, 7659
Vapor Pressure 14mmHg at 20°C
Formula Weight 136.09

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.