missing translation for 'onlineSavingsMsg'
Learn More

Bucetin, TRC

Product Code. 18071150
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18071150 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18071150 Supplier Toronto Research Chemicals Supplier No. TRCB6894201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1083-57-4
Molecular Formula C12 H17 N O3
Molecular Weight (g/mol) 223.2683
Synonym Butanamide, N-(4-ethoxyphenyl)-3-hydroxy-, p-Butyrophenetidide, 3-hydroxy- (6CI,7CI,8CI), N-(4-Ethoxyphenyl)-3-hydroxybutanamide, (±)-Bucetin, 3-Hydroxy-p-butyrophenetidide, Betadid, Bucetin, N-(β-Hydroxybutyryl)-p-phenetidine, p-Ethoxy-N-(β-hydroxybutyryl)aniline, β-Hydroxybutyric acid p-phenetidide
IUPAC Name N-(4-ethoxyphenyl)-3-hydroxybutanamide
SMILES CCOc1ccc(NC(=O)CC(C)O)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H17NO3/c1-3-16-11-6-4-10(5-7-11)13-12(15)8-9(2)14/h4-7,9,14H,3,8H2,1-2H3,(H,13,15)
Formula Weight 223.1208
Chemical Name or Material Bucetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.