missing translation for 'onlineSavingsMsg'
Learn More

Bromperidol, MedChemExpress

Product Code. p-200099878
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448496 100 mg 100mg
16424507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448496 Supplier Medchem Express Supplier No. HYB0901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bromperidol (R-11333) possesses antipsychotic activity, with a high affinity for central dopamine receptors D2. Bromperidol can kill Mycobacteria in a synergistic manner with Spectinomycin.

Bromperidol (R-11333) possesses antipsychotic activity, with a high affinity for central dopamine receptors D2. Bromperidol can kill Mycobacteria in a synergistic manner with Spectinomycin.

Specifications

Chemical Name or Material Bromperidol
CAS 10457-90-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H23BrFNO2
Quantity 100 mg
Synonym R-11333
Solubility Information DMSO : 50 mg/mL (118.96 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(F)C=C1)CCCN2CCC(O)(C3=CC=C(Br)C=C3)CC2
Molecular Weight (g/mol) 420.32
Formula Weight 420.32
Percent Purity 95.72%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.