Learn More
Thermo Scientific™ Oxoid™ Modified Brilliant Green Agar (Dehydrated)

Description
Select and identify salmonellae (other than Salmonella Typhi) from food, feed and water samples with this selective medium.
Select and identify salmonellae (other than Salmonella Typhi) from food, feed and water samples with Thermo Scientific™ Modified Brilliant Green Agar (Dehydrated). The modified formula provides improved inhibition of Escherichia coli and Proteus species. The medium has been widely assessed in Europe and has been used in ISO standards1, 2, 3.
Salmonella are a group of closely related rod-shaped, Gram-negative bacteria. Salmonella typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food. The most common symptoms of Salmonella infection are diarrhoea, fever, abdominal cramps and vomiting.
Use Modified Brilliant Green Agar for selection and identification of salmonellae from food and feed samples.
- Restricts the growth of Pseudomonas species whose colonies may resemble salmonellae on Brilliant Green Agar and cause confusion or extra work to confirm their identity.
- Provides greater inhibition of Escherichia coli and Proteus species than other formulations1.
- ISO compliant: complies with ISO 6579.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Appearance | Light green powder |
| Form | Powder |
| For Use With (Application) | Selective and diagnostic agar for salmonellae (other than Salmonella typhi) from food and feeds |
| pH | 6.7 to 7.1 |
| Quantity | 5 kg |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.