missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BrdU Monoclonal Antibody (MoBU-1)
GREENER_CHOICE

Product Code. 10081378
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10081378 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10081378 Supplier Invitrogen™ Supplier No. B35141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

BrdU Monoclonal Antibody for Flow

Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Flow Cytometry
Classification Monoclonal
Clone MoBU-1
Conjugate Unconjugated
Formulation PBS with 5mM sodium azide; pH 7.2
Gene Alias 5-Bromo-2-deoxyuridine; Bromodeoxyuridine
Host Species Mouse
Immunogen BrdU.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.