Learn More
Description
BRAND ™ Disposable PCR Tubes are designed to deliver exceptional performance for PCR setups, small- and medium-throughput applications, and other workflows requiring precise thermal transfer and high cleanliness. Made from high-quality virgin polypropylene, these tubes ensure reliable results with features that enhance usability and sample integrity. Key features include:
- BIO-CERT™ PCR QUALITY: Certified free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, ensuring the highest level of cleanliness for sensitive PCR applications.
- Superior Heat Transfer: Extra thin-walled tubes optimize thermal transfer during PCR, reducing cycle times and providing consistent performance over a wide temperature range.
- Reliable Sealing Caps: Tight-sealing caps prevent sample evaporation and loss due to heating, while offering easy-to-open and close functionality.
- Color Options: Offered in multiple colors for simplified visual sample tracking and organization.
- High-Quality Construction: Manufactured from high-quality virgin resins without lubricants or chemical additives that could contaminate reagents or samples.
- Autoclavable: Designed to withstand autoclaving at 121°C (2 bar) in compliance with DIN EN 285.
Specifications
Specifications
| Certifications/Compliance | RNase, DNase and DNA free |
| Color | Rose |
| Volume (Metric) | 0.5 mL |
| Quantity | 1000 Pack |
| For Use With (Equipment) | Thermal cyclers for excellent reproducible results in most PCR applications |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Closure Type | Flat Cap |
| Autoclavable | Autoclavable |
| Closure Included | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.