missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Designed for cultivating suspension cell lines where adhesion is undesirable
- Optimized surface reduces cell adhesion and protein adsorption, minimizing enzyme and cellular activation
- Inhibits early differentiation of stem cells
- Orange color code with embossed alphanumeric coding for easy identification
- Made from pure, newly synthesized polystyrene (PS), sterile, and includes lids
- BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
- Manufactured in an ISO Class 7 cleanroom for optimal cleanliness
- Raw materials meet USP and ISO 10993 requirements
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Lid | With Lid |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Packaging | Individually wrapped |
| Color | Transparent |
| Product Type | 96-well microplate |
| Material | Polystyrene |
| Volume (Metric) Well | 350 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?