missing translation for 'onlineSavingsMsg'
Learn More

SCAT Box for Politainer (Foldable Waste Container)

Product Code. 10398719 Shop All SCAT Europe Products
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10398719 20 L Each
10282420 5 L Each
10190330 10 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10398719 Supplier SCAT Supplier No. 107336
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Outer carton for the SCAT Politainer - also known as folding canisters.

SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces

Specifications

Autoclavable Not Autoclavable
Capacity (Metric) 20 L
Color White (Translucent)
Dimensions (L x W x H) 306 x 290 x 290 mm
For Use With (Application) Waste Disposal System
For Use With (Equipment) Politainer
Material Corrugate
Mouth Style Wide
Product Type Cubitainer
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.