Learn More
Invitrogen™ Bovine IL-4 Uncoated ELISA Kit
Description
The Bovine IL-4 Uncoated ELISA Kit contains pre-matched antibody pairs and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of bovine IL-4. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Accession Number | P30367 |
| Assay Range | 15.6 to 1000 pg/mL |
| Assay Sensitivity | <15.6 pg/mL |
| Description | Bovine IL-4 ELISA Reagent Kit |
| Conjugate | HRP |
| Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
| Product Type | ELISA |
| Sample Type | Serum, Supernatant |
| Specificity | No significant cross-reaction with recombinant bovine IL-1ß, IL-2, IL-6, IL-8, IFNg or TNFa; no significant cross-reaction with recombinant human, equine, mouse, porcine, rat, or rhesus macaque IL-4 |
| Sufficient For | 5 x 96 microplate wells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.