missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine IL-1 beta Uncoated ELISA Kit

Product Code. 10178103
Change view
Click to view available options
Quantity:
5 x 96 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10178103 5 x 96 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10178103 Supplier Invitrogen™ Supplier No. ESS0027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Bovine IL-1beta Uncoated ELISA Kit contains pre-matched antibody pairs and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of bovine IL-1beta. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09428
Assay Range 31.3 to 2000 pg/mL
Assay Sensitivity <31.3 pg/mL
Description Bovine IL-1 beta ELISA Reagent Kit
Conjugate HRP
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Product Type ELISA
Sample Type Serum, Supernatant
Specificity No significant cross-reaction with recombinant bovine IL-2, IL-4, IL-6, IL-8, IFNg, or TNFa
Sufficient For 5 x 96 microplate wells
Target Species Bovine
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 281251
Gene Symbol IL1B
Kit Contents Coating Antibody, Lyophilized Recombinant Standard, Detection Antibody, Streptavidin-HRP, Substrate Solution, Ready to use Stop Solution, 0.16M Sulfuric Acid, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
Label or Dye HRP
Measurement Absorbance at 450 nm minus absorbance (at 550 nm)
Precision Not measured
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 100 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Target IL-1 beta
Test Time 24 hrs
Total Assay Time 2 hrs 50 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.