Learn More
Karl Hecht™ Assistent™ Reduction and Expansion Adapters
Description
- Solid rim
- Acc. to DIN 12257
- Reinforced ground joint
Specifications
Specifications
| Cone Size | 24/29 |
| Material | Borosilicate Glass |
| Product Type | Duran sleeve |
| Quantity | 1 Each |
| Standard Taper Joint No. | Socket: 29/32 |